Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CERCAM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565673
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565673 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565673 Supplier Invitrogen™ Supplier No. PA565673
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FDQHPNEQYKA HFWPRDLVAF SAQPLLAAPT HYAGDAEWLS DTETSSPWDD D Highest antigen sequence identity to the following orthologs - mouse 87%, rat 88%.

Probable cell adhesion protein involved in leukocyte transmigration across the blood-brain barrier. Does not express any beta-galactosyltransferase activity in vitro. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CERCAM
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CERCAM
Gene Accession No. Q5T4B2
Gene Alias 2600006K01Rik; AL024097; CEECAM1; CERCAM; cerebral cell adhesion molecule; cerebral endothelial cell adhesion molecule; cerebral endothelial cell adhesion molecule 1; GLT25D3; glycosyltransferase 25 domain containing 3; glycosyltransferase 25 family member 3; Inactive glycosyltransferase 25 family member 3; KIAA1502; Probable inactive glycosyltransferase 25 family member 3
Gene Symbols CERCAM
Host Species Rabbit
Immunogen Recombinant Human CERCAM. Recombinant protein control fragment (Product #RP-106347).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51148
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.