Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERD4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989771
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9771 Supplier Bioss Supplier No. BS13866R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the D4 protein family. The encoded protein is a transcription regulator that binds acetylated histones and is a component of the BAF chromatin remodeling complex. Alternate splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Jul 2013].

Specifications

Antigen CERD4
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene DPF3
Gene Accession No. Q92784
Gene Alias 2810403B03Rik; 6530402L11Rik; Baf45c; BRG1-associated factor 45C; C78788; Cerd4; cer-d4; cerebellum D4; D4 zinc and double PHD finger family 3; D4, zinc and double PHD fingers, family 3; double PHD fingers 3; DPF3; Zinc finger protein cer-d4; Zinc finger protein DPF3
Gene Symbols DPF3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CERD4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8110
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.