Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cerebellin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559498
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559498 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559498 Supplier Invitrogen™ Supplier No. PA559498
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSEPVLLEGE CLVVCEPGRA AAGGPGGAAL GEAPPGRVAF AAVRSHHHEP AGETGNGTSG AIYFDQV Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

Members of the precerebellin family, such as CBLN3, contain a cerebellin motif (see CBLN1; MIM 600432) and a C-terminal C1q signature domain (see MIM 120550) that mediates trimeric assembly of atypical collagen complexes. However, precerebellins do not contain a collagen motif, suggesting that they are not conventional components of the extracellular matrix (Pang et al., 2000 [PubMed 10964938]).[supplied by OMIM, Aug 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cerebellin 3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CBLN3
Gene Accession No. Q6UW01
Gene Alias CBLN3; cerebellin 3 precursor; cerebellin 3 precursor protein; cerebellin-3; precerebellin 3; PRO1486; RGD1563731; UNQ755/PRO1486
Gene Symbols CBLN3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Cerebellin 3. Recombinant protein control fragment (Product #RP-98450).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 643866
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.