Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerebral Protein 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106639
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106639 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106639 Supplier Thermo Scientific Supplier No. PA5106639
Add to Cart
Add to Cart

Cerebral Protein 1 Polyclonal Antibody

Cerebral Protein 1 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cerebral Protein 1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rrp8
Gene Accession No. O43159
Gene Alias 1500003O22Rik; 2900001K19Rik; AW538116; Cerebral protein 1; cerebral protein 1 homolog; HUCE1; hucep1; hucep-1; KIAA0409; NML; Nucleomethylin; RGD1308302; ribosomal RNA processing 8, methyltransferase, homolog (yeast); ribosomal RNA-processing protein 8; Rrp8; RRP-8; RRP8 methyltransferase homolog
Gene Symbols Rrp8
Host Species Rabbit
Immunogen A synthesized peptide derived from human RRP8(Accession O43159), corresponding to amino acid residues F277-R327.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23378
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.