Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cerebral Protein 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536441
Description
This antibody detects endogenous protein at a molecular weight of 51 kDa. Purity is >95% by SDS-PAGE.
Essential component of the eNoSC (energy-dependent nucleolar silencing) complex, a complex that mediates silencing of rDNA in response to intracellular energy status and acts by recruiting histone-modifying enzymes. The eNoSC complex is able to sense the energy status of cell: upon glucose starvation, elevation of NAD(+)/NADP(+) ratio activates SIRT1, leading to histone H3 deacetylation followed by dimethylation of H3 at 'Lys-9' (H3K9me2) by SUV39H1 and the formation of silent chromatin in the rDNA locus. In the complex, RRP8 binds to H3K9me2 and probably acts as a methyltransferase. Its substrates are however unknown.
Specifications
Cerebral Protein 1 | |
Polyclonal | |
Unconjugated | |
Rrp8 | |
1500003O22Rik; 2900001K19Rik; AW538116; Cerebral protein 1; cerebral protein 1 homolog; HUCE1; hucep1; hucep-1; KIAA0409; NML; Nucleomethylin; RGD1308302; ribosomal RNA processing 8, methyltransferase, homolog (yeast); ribosomal RNA-processing protein 8; Rrp8; RRP-8; RRP8 methyltransferase homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
101867, 23378, 308911 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O43159, Q5U4F0, Q9DB85 | |
Rrp8 | |
Synthetic peptide corresponding to amino acids 291-340 of human Cerebral 1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction