Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerebral protein 5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989161
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9161 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9161 Supplier Bioss Supplier No. BS9553R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RTL8C (Retrotransposon Gag Like 8C) is a Protein Coding gene. An important paralog of this gene is RTL8A.

Specifications

Antigen Cerebral protein 5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RTL8C
Gene Accession No. O15255
Gene Alias CAAX box 1; CXX1; FAM127A; family with sequence similarity 127 member A; family with sequence similarity 127, member A; Mammalian retrotransposon derived protein 8C; MAR8; MAR8C; MART8; MART8C; protein FAM127A; Retrotransposon Gag-like protein 8C; RTL8C
Gene Symbols RTL8C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CXX1/Cerebral protein 5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8933
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.