Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceric Ammonium Nitrate, ACS Reagent (Ammonium Cerium (IV) Nitrate), Reagents

Catalog No. 01338055
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-338-055 2 kg
01-338-056 100 g
01-338-054 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-338-055 Supplier Reagents Holdings Llc Supplier No. C11715202A5
Add to Cart
Add to Cart

Ceric Ammonium Nitrate, ACS Reagent (Ammonium Cerium (IV) Nitrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 16774-21-3
Molecular Formula CeH8N8O18
Molecular Weight (g/mol) 548.22
InChI Key WIBGOERAEYJBOT-UHFFFAOYSA-N
Synonym Ammonium Cerium(IV) Nitrate, Ammonium Hexanitratocerate (IV)
IUPAC Name λ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
SMILES N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Orange/Red or Orange/Yellow
pH 1
Physical Form Solid
Quantity 2 kg
Grade ACS
Packaging Poly Bottle
Specific Gravity 2.49
Chemical Name or Material Ceric Ammonium Nitrate, ACS Reagent (Ammonium Cerium (IV) Nitrate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.