Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceric Ammonium Sulfate, 0.100 N (N/10) in 1 N Sulfuric Acid, Ricca Chemical
SDP

Catalog No. 189025 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle

Chemical Identifiers

CAS 10378-47-9, 7732-18-5, 7664-93-9
Molecular Formula CeH20N4O18S4
Molecular Weight (g/mol) 632.53
InChI Key VCNAMBGKEDPVGQ-UHFFFAOYSA-J
Synonym Ceric ammonium sulfate
PubChem CID 16211593
IUPAC Name λ4-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate
SMILES N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O

Specifications

Color Orange
CAS Min % 5.55, 88.0, 4.84
CAS Max % 5.77, 91.0, 5.03
Grade Laboratory
Packaging Cubitainer
Solvent or Matrix 1N Sulfuric Acid
pH <1
Quantity 10 L
Physical Form Liquid
Concentration or Composition Notes 0.100N
Chemical Name or Material Ceric Ammonium Sulfate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.