Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceric Ammonium Sulfate, 0.200 N (N/5), Ricca Chemical
SDP

Catalog No. R18910001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R18910001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R18910001A Supplier Ricca Chemical Company Supplier No. R18910001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9 , 10378-47-9
Molecular Formula CeH20N4O18S4
Molecular Weight (g/mol) 632.53
InChI Key VCNAMBGKEDPVGQ-UHFFFAOYSA-J
Synonym Ceric ammonium sulfate
PubChem CID 16211593
IUPAC Name λ4-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate
SMILES N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O

Specifications

Color Orange
CAS Min % 9.74, 77.0, 11.32
CAS Max % 10.14, 80.0, 11.79
Physical Form Liquid
Quantity 1 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.200 Normal
Packaging Natural Poly Bottle
Chemical Name or Material Ceric Ammonium Sulfate
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.