Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceric Sulfate, 0.250 N (N/4), in approximately 2 N Sulfuric Acid, Ricca Chemical
SDP

Catalog No. 19071 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19071 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19071 Supplier Ricca Chemical Company Supplier No. 19071
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 13590-82-4 , 7664-93-9
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Color Orange
CAS Min % 54.0, 41.49, 2.09
CAS Max % 57.0, 43.19, 2.18
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix approximately 2 Normal Sulfuric Acid
pH <1
Quantity 4 L
Physical Form Liquid
Concentration or Composition Notes 0.250Normal
Chemical Name or Material Ceric Sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.