Learn More
CERILLIANT™ UR-144 4-Hydroxypentyl metabolite, certified reference material, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Worldwide Glass Resource 26636
Description
A certified solution standard suitable for use as starting material in calibrators and controls for GC/MS or LC/MS synthetic cannabinoid testing methods in forensic analysis, clinical toxicology, or urine drug testing. UR-144 5-Hydroxypentyl metabolite is a primary urinary metabolite of UR-144, a third generation synthetic cannabinoid that has seen wide popularity in illicit drug markets since late 2012. This biomarker is also referred to as the terminal omega hydroxyl metabolite of UR-144.Specifications
| 67-56-1 , 1537889-04-5 | |
| MFCD29036719 | |
| 327.46 | |
| 1 mL |
| C21H29NO2 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| Ampule | |
| 9°C (48.2°F) |
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
CERILLIANT is a registered trademark of Cerilliant Corporation