Learn More
Cerium(III) acetate hydrate, 99.995% (metals basis)
Description
Cerium(III) acetate hydrate is used in thin fim buffer layers suitable for high κ dielectrics by sol-gel route. And it is also used as a catalyst for auto-oxidation of cresols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Physical Form | Crystalline |
Quantity | 50 g |
Linear Formula | Ce(C2H3O2)3·xH2O |
Solubility Information | Soluble in water( 3.5g/L). |
Formula Weight | 317.26 (Anhydrous) |
Percent Purity | 99.995% |
Assay | (metals basis) |
Chemical Name or Material | Cerium (III) acetate hydrate |
Safety and Handling
Hazard Category | H315-H318-H335 |
Hazard Statement | GHS H Statement H318-H335-H315 Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.