Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) acetate sesquihydrate, 99.9% (REO)

Catalog No. AA1529436
Click to view available options
Quantity:
100 g
500 g

With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols | CAS: 537-00-8 | C6H9CeO6 | 317.248 g/mol

With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols.Cerium(III) acetate sesquihydrate, acts as a source of cerium involved in the preparation of cerium oxide. It finds application in metallurgy, glass and glass polishing, ceramics, catalysts, and in phosphors. It is utilized in steel production to remove the free sulfur and oxygen. Further, it is an efficient glass polishing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 537-00-8
Molecular Formula C6H9CeO6
Molecular Weight (g/mol) 317.248
MDL Number MFCD00051060
InChI Key VGBWDOLBWVJTRZ-UHFFFAOYSA-K
Synonym cerium acetate, cerous acetate, cerium 3+ acetate, cerium iii acetate, cerium triacetate, cerium 3+ triacetate, unii-yyb5291r2h, acetic acid, cerium 3+ salt, cerium iii acetate sesquihydrate
PubChem CID 10826
IUPAC Name cerium(3+);triacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Ce+3]

Specifications

Melting Point 308°C (decomposition)
Color White
Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water.
Formula Weight 344.28 (317.26 Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Cerium(III) acetate sesquihydrate
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 208-654-0
RTECSNumber FK4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.