Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) ammonium nitrate tetrahydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog Number AA3836736
Click to view available options
Quantity:
100 g
500 g
13083-04-0
CeH16N7O19
558.27
MFCD00213483
AIVZRMAXALRCHB-UHFFFAOYSA-N
ce.3no3.2hno3.4h2o.2nh3, ammonium cerium iii nitrate tetrahydrate, cerous ammonium nitrate, hydrated, reagent, cerium iii ammonium nitrate tetrahydrate, reagent grade, cerium 3+ bis nitric acid diamine tetrahydrate trinitrate
91886599
azane;cerium(3+);nitric acid;trinitrate;tetrahydrate
N.N.O.O.O.O.[Ce+3].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
13083-04-0
CeH16N7O19
558.27
MFCD00213483
AIVZRMAXALRCHB-UHFFFAOYSA-N
ce.3no3.2hno3.4h2o.2nh3, ammonium cerium iii nitrate tetrahydrate, cerous ammonium nitrate, hydrated, reagent, cerium iii ammonium nitrate tetrahydrate, reagent grade, cerium 3+ bis nitric acid diamine tetrahydrate trinitrate
91886599
azane;cerium(3+);nitric acid;trinitrate;tetrahydrate
N.N.O.O.O.O.[Ce+3].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

A useful oxidant for organic synthesis. This chemical is also useful for the synthesis of quinoxaline derivatives which are useful as DNA cleaving agents, dyes, and organic semiconductors. Used to release organic ligands from metal carbonyls. Whereby the metal is oxidized, Carbon monoxide is produced, and the organic ligand is released.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline
Quantity 500 g
Solubility Information Soluble in water.
Formula Weight 558.28 (486.22 Anhydrous)
Grade Reagent
Appearance White crystalline
Chemical Name or Material Cerium(III) ammonium nitrate tetrahydrate, For reagent grade
Hazard Category H272-H315-H319
Precautionary Statement P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit