Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) nitrate hexahydrate, REacton™, 99.5% (REO)

Catalog No. aa1132909
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg

For Separation of Cerium From Other Rare Earths, Catalyst For Hydrolysis of Phosporic Acid Esters | CAS: 10294-41-4 | CeH12N3O15 | 434.22 g/mol

Cerium(III) nitrate hexahydrate is used in the separation of cerium from other rare earths. It plays an important role as a catalyst for hydrolysis of phosphoric acid esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-41-4
Molecular Formula CeH12N3O15
Molecular Weight (g/mol) 434.22
MDL Number MFCD00149631
InChI Key QQZMWMKOWKGPQY-UHFFFAOYSA-N
Synonym cerium iii nitrate hexahydrate, cerium nitrate hexahydrate, cerous nitrate hexahydrate, unii-fl1r38jebb, fl1r38jebb, nitric acid cerium salt, cerium iii nitratehexahydrate, ce.3no3.6h2o, ksc494m5f
PubChem CID 16211466
IUPAC Name cerium(3+) hexahydrate trinitrate
SMILES O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 10 g
Assay Percent Range 99.5%
Solubility Information Soluble in water,alcohol and acetone.
Formula Weight 434.23 (326.13 Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Cerium(III) nitrate hexahydrate, hexahydrate
Hazard Category H272-H315-H318-H350
Precautionary Statement P201-P202-P220-P221-P264b-P271-P280-P302+P352-P305+P351+P338-P308+P313-P310-P332+P313-P362-P370+P380+P375-P372-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 233-297-2
RTECSNumber FK6300000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.