Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) phosphate, 99% min

Catalog No. p-7050654
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 13454-71-2 | CeO4P | 235.09 g/mol

Intermediate product in cerium-based technique for staining lysosomal enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-71-2
Molecular Formula CeO4P
Molecular Weight (g/mol) 235.09
MDL Number MFCD00049424
InChI Key TYAVIWGEVOBWDZ-UHFFFAOYSA-K
Synonym cerium phosphate, cerium 3+ phosphate, unii-xr1z4biw2j, cerium iii phosphate, xr1z4biw2j, phosphoric acid,cerium 3+ salt 1:1, cerium monophosphate, cerium 3 phosphate, cerium orthophosphate cepo4, acmc-20alhk
PubChem CID 159673
IUPAC Name cerium(3+);phosphate
SMILES [Ce+3].[O-]P([O-])([O-])=O

Specifications

Color Yellow
Physical Form Powder
Quantity 25 g
Linear Formula CePO4
Solubility Information Insoluble in water.
Formula Weight 235.09
Percent Purity ≤99%
Odor Odorless
Density 5.22 g/mL
Chemical Name or Material Cerium(III) phosphate
EINECSNumber 236-637-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.