Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) sulfide, 99.9% (metals basis)

Catalog No. p-4407354
Click to view available options
Quantity:
5 g
25 g

CAS: 12014-93-6 | Ce2S3 | 376.41 g/mol

Used as pharmaceutical intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12014-93-6
Molecular Formula Ce2S3
Molecular Weight (g/mol) 376.41
MDL Number MFCD00016007
InChI Key MMXSKTNPRXHINM-UHFFFAOYSA-N
Synonym cerium sulfide, dicerium trisulphide, ceric sulfide, cerium 3+ trisulfide, dicerium 3+ trisulfide, dicerium 3+ trisulfanediide
PubChem CID 159394
SMILES [S--].[S--].[S--].[Ce+3].[Ce+3]

Specifications

Melting Point 2100°C
Color Red to Brown
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Insoluble in water.
IUPAC Name cerium(3+);trisulfide
Formula Weight 376.43
Odor Rotten eggs
Appearance Red-brown powder
Density 5.02 g/cm3 at 20°C
Chemical Name or Material Cerium(III) sulfide
Show More Show Less
Hazard Category H261-H315-H319-H335
Precautionary Statement P231+P232-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation DOT Class: 4.3, Packaging Group: III
EINECSNumber 234-603-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.