Alfa Aesar™ Cerium(IV) isopropoxide, Ce 37-45%

Manufacturer: Alfa Aesar™ 4446806

Quantity 5g

 View more versions of this product

Catalog No. AA4446806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 63007-83-0
    Molecular Formula C12H28CeO4
    Linear Formula Ce(OC3H7)4
    Formula Weight 376.48
    MDL Number MFCD00061483
    UN Number UN2925
    Chemical Name or Material Cerium (IV) isopropoxide

    Specifications

    Formula Weight 376.48
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Molecular Formula C12H28CeO4
    Linear Formula Ce(OC3H7)4
    CAS 63007-83-0
    UN Number UN2925
    Solubility Information Soluble in pyridine
    Sensitivity Air & Moisture Sensitive
    Physical Form Powder
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H228
    Causes severe skin burns and eye damage.
    Flammable solid.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    Color Orange-red to yellow
    MDL Number MFCD00061483
    Concentration Ce 37 to 45%
    Melting Point 140°C (dec.)
    Quantity 5g
    Chemical Name or Material Cerium (IV) isopropoxide

    GHS H Statement
    H314-H228
    Causes severe skin burns and eye damage.
    Flammable solid.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit