Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(IV) oxide, 20% in H2O, colloidal dispersion in water with dispersant, NanoTek™, Thermo Scientific™

Catalog No. AA4491022
Click to view available options
:
100g

Catalog No. AA4491022

Supplier: Thermo Scientific Chemicals 4491022

For Use As a Cerium Oxide Water Borne Dispersion Suitable For Use in Alkaline Side Polishing Slurries. Submicron Particle Size Makes For Good Removal Rate With Minimal Subsurface Damage

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1306-38-3
Quantity 100g
Molecular Formula CeO2
MDL Number MFCD00010933
Synonym ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
InChI Key CETPSERCERDGAM-UHFFFAOYSA-N
SMILES O=[Ce]=O
IUPAC Name dioxocerium
Molecular Weight (g/mol) 172.114
PubChem CID 73963
ChEBI CHEBI:79089
Formula Weight 172.12
Chemical Name or Material Cerium(IV) oxide, CE-6080, 20% in H2O, colloidal dispersion with dispersant
Show More Show Less
Hazard Category H303 : May be harmful if swallowed.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.