Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(IV) sulfate, anhydrous, 97%

Catalog No. AAL2025436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2025436 500 g
AAL2025414 25 g
AAL2025422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL2025436 Supplier Thermo Scientific Chemicals Supplier No. L2025436
Add to Cart

CAS: 13590-82-4 | CeO8S2 | 332.23 g/mol

Cerium(IV) sulfate is a redox indicator and is used in analytical chemistry for redox titrations. In ionized form, ceric ion acts as a strong oxidizer under acidic conditions. It is also used to prepare spherical-like nanoparticles CeO2, nanoparticles for NO reductions' reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerium(IV) sulfate is a redox indicator and is used in analytical chemistry for redox titrations. In ionized form, ceric ion acts as a strong oxidizer under acidic conditions. It is also used to prepare spherical-like nanoparticles CeO2, nanoparticles for NO reductions′ reactions.

Solubility
Soluble in water and dilute sulfuric acid.

Notes
Store in cool place. Hygroscopic and moisture sensitive. Avoid moisture. Incompatible with strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13590-82-4
Molecular Formula CeO8S2
Molecular Weight (g/mol) 332.23
MDL Number MFCD00148852
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
Synonym ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate
PubChem CID 159684
IUPAC Name cerium(4+);disulfate
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Quantity 500 g
UN Number UN3085
Merck Index 14,1990
Solubility Information Soluble in water and dilute sulfuric acid.
Formula Weight 332.24
Percent Purity 97%
Sensitivity Hygroscopic
Density 3.91
Chemical Name or Material Cerium(IV) sulfate, anhydrous
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 237-029-5
RTECSNumber WS6960000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.