Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(IV) sulfate hydrate, REacton™, 99% (REO)

Catalog No. p-7048718
Click to view available options
Quantity:
100 g
500 g

CAS: 95838-16-7 | CeO8S2 | 332.23 g/mol

Cerium(IV) sulfate hydrate acts as oxidizing agent which is used for the generation of o-benzoquinone from catechol. It promotes the formation of radicals from dialkyl malonates, which undergo substitution reactions with furans and thiophenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95838-16-7
Molecular Formula CeO8S2
Molecular Weight (g/mol) 332.23
MDL Number MFCD00148852
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
Synonym sulfuricacid, cerium salt, hydrate 9ci, acmc-20mqih, cerium iv sulfate monohydrate, cerium iv sulfate hydrate, reacton, cerium 4+ sulfate-water 1/2/1
PubChem CID 20223355
IUPAC Name λ⁴-cerium(4+) hydrate disulfate
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 195°C (decomposition)
Physical Form Crystalline
Quantity 100 g
UN Number UN3085
Solubility Information Slightly soluble in water. Soluble in dil.sulfuric acid.
Formula Weight 332.24 (Anhydrous)
Percent Purity 99%
Grade Reacton™
Sensitivity Moisture sensitive
Density 3.01 g/mL
Assay (REO)
Chemical Name or Material Cerium(IV) sulfate hydrate
Show More Show Less
Hazard Category H272-H314-H335
Hazard Statement GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire
oxidizer.
Precautionary Statement P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.
EINECSNumber 237-029-5
RTECSNumber WS6960000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.