Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(IV) sulfate tetrahydrate, 98+%, Acros Organics

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
10294-42-5
CeO8S2·4H2O
404.29
MFCD00149427
MEXSQFDSPVYJOM-UHFFFAOYSA-J
cerium iv sulfate tetrahydrate,ceric sulfate tetrahydrate,unii-6sax34b8je,6sax34b8je,acmc-20ecws,ceric sulphate tetrahydrate,ksc493s0d,aronis24302,sulfuric acid, cerium 4+ salt 2:1 , tetrahydrate,cerium iv sulfate hydrate
22723557
cerium(4+);disulfate;tetrahydrate
O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+4]
10294-42-5
CeO8S2·4H2O
404.29
MFCD00149427
MEXSQFDSPVYJOM-UHFFFAOYSA-J
cerium iv sulfate tetrahydrate,ceric sulfate tetrahydrate,unii-6sax34b8je,6sax34b8je,acmc-20ecws,ceric sulphate tetrahydrate,ksc493s0d,aronis24302,sulfuric acid, cerium 4+ salt 2:1 , tetrahydrate,cerium iv sulfate hydrate
22723557
cerium(4+);disulfate;tetrahydrate
O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+4]
Aluminum (Al) 1000ppm max.
Assay Percent Range 98% min. (Iodometry)
Linear Formula Ce(SO4)2·4H2O
Fieser 02,65; 15,73
Merck Index 14, 1990
Solubility Information Solubility in water: 38g/L (50°C). Other solubilities: soluble in dilute sulfuric acid,slowly soluble in cold,more readily soluble in,hot mineral acids
Formula Weight 404.29
Physical Form Powder
Percent Purity 98+%
Chloride 100ppm max
Chlorine (Cl) 100ppm max.
Copper (Cu) 30ppm max.
Density 1.06
Heavy Metals (as Pb) 30ppm max.
Insoluble Matter 0.1% max. (in H2SO4)
Iron (Fe) 150ppm max.
Lead (Pb) 30ppm max.
Nickel (Ni) 30ppm max.
Packaging Plastic bottle
Substances not ppt by NH4OH 0.3% max.
Color Yellow
Quantity 100g
Chemical Name or Material Cerium(IV) sulfate tetrahydrate, 99+%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger