Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CERK Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01950191
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-950-191 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-950-191 Supplier Invitrogen™ Supplier No. PA116963
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: A549 lysate, antigen standard for CERK (transient overexpression lysate).

CERK converts ceramide to ceramide 1-phosphate (C1P), a sphingolipid metabolite. Both CERK and C1P have been implicated in various cellular processes, including proliferation, apoptosis, phagocytosis, and inflammation (Kim et al., 2006 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CERK
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% sodium azide
Gene CERK
Gene Accession No. Q8K4Q7, Q8TCT0
Gene Alias acylsphingosine kinase; AI848610; Ceramide kinase; Cerk; CERK antibody; D330016D08Rik; dA59H18.2; dA59H18.3; hCERK; KIAA1646; Lipid kinase 4; lipid kinase LK4; LK4; mCERK; mKIAA1646
Gene Symbols CERK
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of human Ceramide Kinase (between residues 50-150).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223753, 64781
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.