Learn More
Invitrogen™ CERK Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532707
Description
Percent identity with other species by BLAST analysis: Human (100%); Gorilla, Gibbon, Monkey, Marmoset (90%); Dog, Panda (85%); Bovine, Elephant, Rabbit (80%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
CERK converts ceramide to ceramide 1-phosphate (C1P), a sphingolipid metabolite. Both CERK and C1P have been implicated in various cellular processes, including proliferation, apoptosis, phagocytosis, and inflammation (Kim et al., 2006 ).
Specifications
| CERK | |
| Polyclonal | |
| Unconjugated | |
| CERK | |
| acylsphingosine kinase; AI848610; Ceramide kinase; Cerk; CERK antibody; D330016D08Rik; dA59H18.2; dA59H18.3; hCERK; KIAA1646; Lipid kinase 4; lipid kinase LK4; LK4; mCERK; mKIAA1646 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64781 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q8TCT0 | |
| CERK | |
| Synthetic 20 amino acid peptide from N-terminal extracellular domain of human CERK. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.