Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CERK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA513856
Description
CERK Polyclonal Antibody for Western Blot
CERK converts ceramide to ceramide 1-phosphate (C1P), a sphingolipid metabolite. Both CERK and C1P have been implicated in various cellular processes, including proliferation, apoptosis, phagocytosis, and inflammation (Kim et al., 2006 ).
Specifications
CERK | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8K4Q7, Q8TCT0 | |
CERK | |
KLH conjugated synthetic peptide between 487-516 amino acids from the C-terminal region of human CERK. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CERK | |
acylsphingosine kinase; AI848610; Ceramide kinase; Cerk; CERK antibody; D330016D08Rik; dA59H18.2; dA59H18.3; hCERK; KIAA1646; Lipid kinase 4; lipid kinase LK4; LK4; mCERK; mKIAA1646 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
223753, 64781 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction