Learn More
Invitrogen™ CERKL Polyclonal Antibody

Description
CERKL Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Specifications
Specifications
| Antigen | CERKL |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 3.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | CERKL |
| Gene Accession No. | Q49MI3 |
| Gene Alias | ceramide kinase like; ceramide kinase-like; ceramide kinase-like protein; CERKL; Gm1958; retinitis pigmentosa 26 (autosomal recessive); Rp26; zgc:162213 |
| Gene Symbols | CERKL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.