Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerotic Acid 95.0+%, TCI America™
SDP

Catalog No. C082910G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C082910G 10 g
C08291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C082910G Supplier TCI America Supplier No. C082910G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 506-46-7
Molecular Formula C26H52O2
Molecular Weight (g/mol) 396.7
MDL Number MFCD00002811
InChI Key XMHIUKTWLZUKEX-UHFFFAOYSA-N
Synonym cerotic acid, ceric acid, ceratinic acid, cerinic acid, hexacosanic acid, cerylic acid, cerotinic acid, n-hexacosanoic acid, unii-d42cqn6p36, cerate
PubChem CID 10469
ChEBI CHEBI:31009
IUPAC Name hexacosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 88°C
Color White-Yellow
Quantity 10 g
Formula Weight 396.70
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cerotic Acid
EINECSNumber (2)-0608
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.