Learn More
CERS2 Mouse anti-Human, Clone: OTI3D9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809918

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
LASS2 is a protein that has sequence similarity to yeast longevity assurance gene 1. Mutation or overexpression of the related gene in yeast has been shown to alter yeast lifespan. The human protein may play a role in the regulation of cell growth. This gene encodes a protein that has sequence similarity to yeast longevity assurance gene 1. Mutation or overexpression of the related gene in yeast has been shown to alter yeast lifespan. The human protein may play a role in the regulation of cell growth. Alternatively spliced transcript variants encoding the same protein have been described.Specifications
CERS2 | |
Monoclonal | |
Unconjugated | |
CERS2 | |
L3, LASS2, SP260, TMSG1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
29956 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3D9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96G23 | |
CERS2 | |
Human recombit protein fragment corresponding to amino acids 62-136 of human CERS2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.