Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CERT Monoclonal Antibody (04)
GREENER_CHOICE

Catalog No. PIMA530261
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530261 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530261 Supplier Invitrogen™ Supplier No. MA530261
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human COL4A3BP.

CERT (GPBP) is a GC-rich promotor-specific transactivating transcription factor. This protein is suggested to play a role in the development of atherosclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CERT
Applications ELISA
Classification Monoclonal
Clone 04
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CERT1
Gene Accession No. Q9Y5P4
Gene Alias 2810404O15Rik; 9230101K08Rik; alpha 3 type IV collagen binding protein; AU016711; Ceramide transfer protein; ceramide transport protein; ceramide transporter; Cert; CERTL; Col4a3bp; collagen type IV alpha 3 binding protein; collagen type IV alpha-3-binding protein; collagen, type IV, alpha 3 (Goodpasture antigen) binding protein; goodpasture antigen-binding protein; GPBP; hCERT; lipid-transfer protein CERTL; MRD34; procollagen, type IV, alpha 3 (Goodpasture antigen) binding protein; STARD11; StAR-related lipid transfer (START) domain containing 11; stAR-related lipid transfer protein 11; START domain-containing protein 11
Gene Symbols CERT1
Host Species Mouse
Immunogen Recombinant Human COL4A3BP protein (Met1-Phe598).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10087
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.