Learn More
MilliporeSigma™ Supelco™ Certified Polypropylene Bonded Caps with Septa
Click to view available options
Septum:
Bonded/Silicone
Rubber
Type:
Hole Cap
Hole Cap (bonded septa)
Description
- Manufactured using virgin resins to eliminate the possibility of sample contamination.
- Caps are molded to ensure consistency in fit, thread specification and overall dimensions.
- The PTFE/silicone septa are manufactured, conditioned and then tested by GC/MS to ensure they have the lowest level of extractable in the industry.
Specifications
Specifications
Diameter (Metric) Cap | 9 mm |
Color | Dark Blue |
Material | PTFE, Polypropylene |
Pre-slit | Yes |
Septum | Bonded/Silicone |
No. per Pack | 100 |
Type | Hole Cap (bonded septa) |