Learn More
Mettler Toledo™ Certified Temperature Kits
Catalog No. p-7193697 Shop All METTLER TOLEDO Products
Click to view available options
For Use With (Equipment):
HE73, HE53 Moisture Analyzer
HX204, HS153, HC103 Moisture Analyzer
Quick and easy temperature testing, for use with Mettler Toledo™ Moisture Analyzers
  • Quick and easy temperature testing
  • Adjusts instrument at the selected operating temperature to ensure reliable results
  • For use with Mettler Toledo™ Moisture Analyzers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) HE73, HE53 Moisture Analyzer
Type Temperature Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.