Learn More
MilliporeSigma™ Certipur™ Refractive Index Standard Kits
Shop All MilliporeSigma Products
Click to view available options
Type:
2,2,4-Trimethyl-pentane/Water
Toluene/Water
Description
These standards are for the calibration of refractometers. We offer different refractive index standards in order to perform calibration in the same range as the final measurement.
- Stable solutions with minimum shelf life of 3 years
- Ready-to-use solutions
- Each standard kit is packed with a certificate of analysis
- Applicable at 20°C and 25°C measuring temperature
- According to Ph Eur. 2.2.6
Specifications
Specifications
| Certifications/Compliance | Traceable to SRM from NIST and PTB n (20°C)=1,4969 Certipur™ |
| Type | Toluene/Water |