Learn More
Invitrogen™ Ceruloplasmin Polyclonal Antibody

Description
Store as a concentrated solution. Centrifuge briefly prior to opening vial.
Specifications
Specifications
| Antigen | Ceruloplasmin |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.3 |
| Gene | CP |
| Gene Accession No. | P00450 |
| Gene Alias | bilitranslocase; CERP; Ceruloplasmin; ceruloplasmin (ferroxidase); CP; CP-2; D3Ertd555e; ferroxidase |
| Gene Symbols | CP |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.