Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ceruloplasmin Recombinant Rabbit Monoclonal Antibody (6C3K9)
GREENER_CHOICE

Catalog No. PIMA538035
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538035 100 μL
MA568575 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA538035 Supplier Invitrogen™ Supplier No. MA538035
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Mouse serum, Rat testis, Rat serum Immunogen sequence: IRGKHVRHYY IAAEEIIWNY APSGIDIFTK ENLTAPGSDS AVFFEQGTTR IGGSYKKLVY REYTDASFTN RKERGPEEEH LGILGPVIWA EVGDTIRVTF HNKGAYPLSI EPIGVRFNKN NEGTYYSPNY NPQSRSVPPS ASHVAPTETF TYEWTVPKEV GPTNADPVCL AKMYYSAVDP TKDIFTGLIG PMKICKKGSL HANGRQK.

Ceruloplasmin encodes a protein that is a metalloprotein that binds most of the copper in plasma and is involved in the peroxidation of Fe(II)transferrin to Fe(III) transferrin. Mutations in this gene cause aceruloplasminemia, which results in iron accumulation and tissue damage, and is associated with diabetes and neurologic abnormalities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ceruloplasmin
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 6C3K9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.05% ProClin 300; pH 7.3
Gene CP
Gene Accession No. P00450, P13635, Q61147
Gene Alias bilitranslocase; CERP; Ceruloplasmin; ceruloplasmin (ferroxidase); CP; CP-2; D3Ertd555e; ferroxidase
Gene Symbols CP
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12870, 1356, 24268
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.