Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CES1 Monoclonal Antibody (3F10)
GREENER_CHOICE

Catalog No. PIMA549209
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549209 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549209 Supplier Invitrogen™ Supplier No. MA549209
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human THP-1 whole cell, human HEPG2 whole cell, rat liver tissue, mouse liver tissue. IHC: human liver cancer tissue, human tonsil tissue, human prostate cancer tissue, mouse lung tissue, rat liver tissue. ICC/IF: HEPG2 cell. Flow: HEPG2 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This gene encodes a member of the carboxylesterase large family. The family members are responsible for the hydrolysis or transesterification of various xenobiotics, such as cocaine and heroin, and endogenous substrates with ester, thioester, or amide bonds. They may participate in fatty acyl and cholesterol ester metabolism, and may play a role in the blood-brain barrier system. This enzyme is the major liver enzyme and functions in liver drug clearance. Mutations of this gene cause carboxylesterase 1 deficiency. Three transcript variants encoding three different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CES1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3F10
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene CES1
Gene Accession No. P10959, P23141, P23953, Q63108, Q64176, Q8VCC2
Gene Alias ACAT; acyl coenzyme A:cholesterol acyltransferase; acyl-coenzyme A:cholesterol acyltransferase; Brain carboxylesterase hBr1; carboxyesterase ES-3; carboxylesterase 1; carboxylesterase 1 (monocyte/macrophage serine esterase 1); carboxylesterase 1A; carboxylesterase 1E; Carboxylesterase 1G; carboxylesterase 1-like; carboxylesterase 2 (liver); CE-1; CEH; Ces1; Ces-1; Ces1a; Ces1e; Ces1g; Ces1l; CES2; cholesteryl ester hydrolase; cocaine carboxylesterase; Eg; EG244595; Egasyn; Es22; Es-22; ES-HTEL; esterase 22; esterase-22; ES-x; Gm4976; hCE-1; HMSE; HMSE1; human monocyte/macrophage serine esterase 1; liver carboxylesterase 1; Liver carboxylesterase 22; liver carboxylesterase 3; methylumbelliferyl-acetate deacetylase 1; MGC117365; Monocyte/macrophage serine esterase; PCE-1; pI 5.5 esterase; REH; Retinyl ester hydrolase; serine esterase 1; SES1; Ses-1; TGH; triacylglycero; Triacylglycerol hydrolase
Gene Symbols CES1, Ces1a, Ces1c, Ces1e, Ces1g
Host Species Mouse
Immunogen E. coli-derived human Liver Carboxylesterase 1/CES1 recombinant protein (Position: E99-A206).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1066, 12623, 13884, 13897, 24346, 244595, 29225, 679817
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.