Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CES1 Recombinant Rabbit Monoclonal Antibody (7C9R6)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535117
Description
Immunogen sequence: MWLRAFILAT LSASAAWGHP SSPPVVDTVH GKVLGKFVSL EGFAQPVAIF LGIPFAKPPL GPLRFTPPQP AEPWSFVKNA TSYPPMCTQD PKAGQLLSEL.
This gene encodes a member of the carboxylesterase large family. The family members are responsible for the hydrolysis or transesterification of various xenobiotics, such as cocaine and heroin, and endogenous substrates with ester, thioester, or amide bonds. They may participate in fatty acyl and cholesterol ester metabolism, and may play a role in the blood-brain barrier system. This enzyme is the major liver enzyme and functions in liver drug clearance. Mutations of this gene cause carboxylesterase 1 deficiency. Three transcript variants encoding three different isoforms have been found for this gene.
Specifications
CES1 | |
Recombinant Monoclonal | |
0.8 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P10959, P23141, P23953, Q63108, Q64176, Q8VCC2 | |
CES1, Ces1a, Ces1c, Ces1e, Ces1g | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CES1 (P23141). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot, Immunocytochemistry | |
7C9R6 | |
Unconjugated | |
CES1 | |
ACAT; acyl coenzyme A:cholesterol acyltransferase; acyl-coenzyme A:cholesterol acyltransferase; Brain carboxylesterase hBr1; carboxyesterase ES-3; carboxylesterase 1; carboxylesterase 1 (monocyte/macrophage serine esterase 1); carboxylesterase 1A; carboxylesterase 1E; Carboxylesterase 1G; carboxylesterase 1-like; carboxylesterase 2 (liver); CE-1; CEH; Ces1; Ces-1; Ces1a; Ces1e; Ces1g; Ces1l; CES2; cholesteryl ester hydrolase; cocaine carboxylesterase; Eg; EG244595; Egasyn; Es22; Es-22; ES-HTEL; esterase 22; esterase-22; ES-x; Gm4976; hCE-1; HMSE; HMSE1; human monocyte/macrophage serine esterase 1; liver carboxylesterase 1; Liver carboxylesterase 22; liver carboxylesterase 3; methylumbelliferyl-acetate deacetylase 1; MGC117365; Monocyte/macrophage serine esterase; PCE-1; pI 5.5 esterase; REH; Retinyl ester hydrolase; serine esterase 1; SES1; Ses-1; TGH; triacylglycero; Triacylglycerol hydrolase | |
Rabbit | |
Affinity chromatography | |
RUO | |
1066, 12623, 13884, 13897, 24346, 244595, 29225, 679817 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction