Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CES1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | CES1 |
---|---|
Concentration | 0.22 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1253
![]() ![]() |
Proteintech Group Inc
169121AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a member of the carboxylesterase large family. The family members are responsible for the hydrolysis or transesterification of various xenobiotics, such as cocaine and heroin, and endogenous substrates with ester, thioester, or amide bonds. They may participate in fatty acyl and cholesterol ester metabolism, and may play a role in the blood-brain barrier system. This enzyme is the major liver enzyme and functions in liver drug clearance. Mutations of this gene cause carboxylesterase 1 deficiency. Three transcript variants encoding three different isoforms have been found for this gene.Specifications
CES1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P23141 | |
1066 | |
CES1 Fusion Protein Ag10567 | |
Primary | |
-20°C | |
CES1 |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ACAT, Brain carboxylesterase hBr1, CE1, CEH, CES1, CES2, Cocaine carboxylesterase, Egasyn, HMSE, HMSE1, Liver carboxylesterase 1, PCE 1, REH, Retinyl ester hydrolase, Serine esterase 1, SES1, TGH, Triacylglycerol hydrolase | |
CES1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title