Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CES2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547715
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547715 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547715 Supplier Invitrogen™ Supplier No. PA547715
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 30% cross-reactivity with recombinant human CES2 is observed and approximately 5% cross-reactivity with recombinant mouse (rm) CES3 and rmCES5 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

CES2 is a member of the carboxylesterase large family. The family members are responsible for the hydrolysis or transesterification of various xenobiotics, such as cocaine and heroin, and endogenous substrates with ester, thioester, or amide bonds. They may participate in fatty acyl and cholesterol ester metabolism, and may play a role in the blood-brain barrier system. The protein encoded by this gene is the major intestinal enzyme and functions in intestine drug clearance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CES2
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Ces2e
Gene Accession No. Q8BK48, Q91WG0
Gene Alias 9030624L02Rik; 9130231C15Rik; ACH M1; Acylcarnitine hydrolase; acylcarnitine hydrolase M1; AI266984; Carboxylesterase 2; carboxylesterase 2 (intestine, liver); carboxylesterase 2A; carboxylesterase 2C; carboxylesterase 2E; carboxylesterase 5; carboxylesterase 6; carboxylesterase isoenzyme; carboxylesterase ML3; CE-2; CES 2; Ces2; CES-2; Ces2a; CES2A1; ces2A3; Ces2c; Ces2e; Ces2l; Ces5; Ces6; CESRL4; Cocaine esterase; hCE-2; ICE; intestinal carboxylesterase; liver carboxylesterase-2; methylumbelliferyl-acetate deacetylase 2; P iCE; PCE-2; Peroxisome proliferator-inducible acylcarnitine hydrolase; phenobarbital-inducible carboxylesterase (liver); pyrethroid hydrolase Ces2a; Pyrethroid hydrolase Ces2e
Gene Symbols Ces2c, Ces2e
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Carboxylesterase 2/CES2 Gln27-Lys557 (predicted).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234671, 234673
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.