Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CES3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547802
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547802 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547802 Supplier Invitrogen™ Supplier No. PA547802
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody detects recombinant mouse Carboxylesterase 3/CES3 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) CES1, rhCES2, recombinant mouse (rm) CES2, and rmCES5 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

CES3 major lipase in white adipose tissue. Involved in the metabolism of xenobiotics and of natural substrates. Hydrolyzes triacylglycerols and monoacylglycerols, with a preference for monoacylglycerols. The susceptibility of the substrate increases with decreasing acyl chain length of the fatty acid moiety. Catalyzes the synthesis of fatty acid ethyl esters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CES3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Ces3a
Gene Accession No. P16303, Q63880, Q8VCT4
Gene Alias Carboxyesterase ES-10; carboxylesterase 1D; Carboxylesterase 3; carboxylesterase 3 (brain); carboxylesterase 3, pseudogene 1; carboxylesterase 3A; Ces1; Ces1d; CES3; Ces31; Ces3a; Ces3-ps1; EC 3.1.1; EC 3.1.1.1; ES31; ES31FLJ21736; ES-HVEL; ES-male; Es-male carboxylesterase; esterase 31; esterase-31; FAEE synthase; fatty acid ethyl ester synthase; Liver carboxylesterase 10; liver carboxylesterase 31; Liver carboxylesterase 31 homolog; pI 6.1 esterase; TGH; Triacylglycerol hydrolase; UNQ869/PRO1887
Gene Symbols Ces1d, Ces3a
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Carboxylesterase3/CES3 Tyr19-Glu561.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104158, 113902, 382053, 689754
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.