Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CES3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592252
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592252 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592252 Supplier Invitrogen™ Supplier No. PA592252
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WPLAQKIANT LACSSSSPAE MVQCLQQKEG EELVLSKKLK NTIYPLTVDG TVFPKSPKEL LKEKPFHSVP FLMGVNNHEF SWLIPRGWGL LDTMEQMSRE DMLAISTPVL TSLDVPPEMM PTVIDEYLGS NSDAQAKCQA FQEFMGDVFI NVPTVSFSRY LRDSGSPVFF YEFQHRPSSF AKIKPAWVKA DHGAEGAFVF GGPFLMDESS RLEATEEEKQ LSLTMMAQWT HFARTGDPNS KALPPWPQFN QAEQYLEINP VPRAGQKFRE AWMQFWSETL PSKIQQWHQK QKNRKAQEDL; Positive Samples: LO2, HT-29, Mouse small intestine, Mouse large intestine, Rat liver, Rat small intestine; Cellular Location: Endoplasmic reticulum lumen.

CES3 major lipase in white adipose tissue. Involved in the metabolism of xenobiotics and of natural substrates. Hydrolyzes triacylglycerols and monoacylglycerols, with a preference for monoacylglycerols. The susceptibility of the substrate increases with decreasing acyl chain length of the fatty acid moiety. Catalyzes the synthesis of fatty acid ethyl esters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CES3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CES3
Gene Accession No. P16303, Q63880, Q6UWW8, Q8VCT4
Gene Alias Carboxyesterase ES-10; carboxylesterase 1D; Carboxylesterase 3; carboxylesterase 3 (brain); carboxylesterase 3, pseudogene 1; carboxylesterase 3A; Ces1; Ces1d; CES3; Ces31; Ces3a; Ces3-ps1; EC 3.1.1; EC 3.1.1.1; ES31; ES31FLJ21736; ES-HVEL; ES-male; Es-male carboxylesterase; esterase 31; esterase-31; FAEE synthase; fatty acid ethyl ester synthase; Liver carboxylesterase 10; liver carboxylesterase 31; Liver carboxylesterase 31 homolog; pI 6.1 esterase; TGH; Triacylglycerol hydrolase; UNQ869/PRO1887
Gene Symbols Ces1d, CES3, Ces3a
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 269-568 of human CES3 (NP_0011721061).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104158, 113902, 23491, 382053, 689754
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.