Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CES6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568885
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568885 Supplier Invitrogen™ Supplier No. PA568885
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Mouse: 100%; Rat: 100%.

Carboxylesterases that catalyzes the hydrolysis of pyrethroids pesticides. Hydrolyzes permethrin faster than cypermethrin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CES6
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ces2a
Gene Accession No. Q8QZR3
Gene Alias 9130231C15Rik; AI266984; carboxylesterase 2A; carboxylesterase 6; carboxylesterase isoenzyme; Ces2a; Ces6; Pyrethroid hydrolase Ces2a
Gene Symbols Ces2a
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse CES6 (aa 251-300).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102022
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.