Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CES7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112340
Description
This product can be used in direct ELISA.
CES7 is involved in the detoxification of xenobiotics and in the activation of ester and amide prodrugs.
Specifications
| CES7 | |
| Polyclonal | |
| PBS with no preservative | |
| Q6AW46 | |
| CES5A | |
| Recombinant Mouse CES5/Carboxylesterase 5 Protein. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| CES5A | |
| 1700081L16Rik; 1700122C07Rik; BB081581; carboxylesterase 5; carboxylesterase 5A; carboxylesterase 7; carboxylesterase-like urinary excreted protein homolog; CAUXIN; CES4C1; CES5; Ces5a; CES7; epididymis luminal protein 126; Gm503; HEL126 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 67935 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction