Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CESA6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144511
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144511 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144511 Supplier Invitrogen™ Supplier No. PA5144511
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CESA6 Polyclonal Antibody for Western Blot, ELISA

Probable catalytic subunit of cellulose synthase terminal complexes ('rosettes'), required for beta-1,4-glucan microfibril crystallization, a major mechanism of the cell wall formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CESA6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene CESA6
Gene Accession No. Q6YVM4
Gene Alias catalytic subunit 6; Os07g0252400
Gene Symbols CESA6
Host Species Rabbit
Immunogen Recombinant Oryza sativa subsp. japonica (Rice) CESA6 protein (124-280aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4342850
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.