Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium carbonate, 99.9% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
5g
25g
100g
500g
534-17-8
CCs2O3
325.819
MFCD00010957
FJDQFPXHSGXQBY-UHFFFAOYSA-L
cesium carbonate,dicesium carbonate,caesium carbonate,cesiumcarbonate,carbonic acid, dicesium salt,cs2co3,unii-qqi20a14p4,cesium carbonate cs2co3,carbonic acid, cesium salt 1:2,dicaesium 1+ ion carbonate
10796
dicesium;carbonate
C(=O)([O-])[O-].[Cs+].[Cs+]
534-17-8
CCs2O3
325.819
MFCD00010957
FJDQFPXHSGXQBY-UHFFFAOYSA-L
cesium carbonate,dicesium carbonate,caesium carbonate,cesiumcarbonate,carbonic acid, dicesium salt,cs2co3,unii-qqi20a14p4,cesium carbonate cs2co3,carbonic acid, cesium salt 1:2,dicaesium 1+ ion carbonate
10796
dicesium;carbonate
C(=O)([O-])[O-].[Cs+].[Cs+]
Assay Percent Range 99.9% (metals basis)
Beilstein 4546405
Solubility Information Freely soluble in water. Soluble in ethanol,dimethylformamide and dimethyl sulfoxide.
Formula Weight 325.82
Physical Form -20 Mesh Powder/Granules
Density 4.072
Sensitivity Hygroscopic
Melting Point 610°C (decomposition)
Quantity 5g
Chemical Name or Material Cesium carbonate

P201-P202-P260-P281-P305+P351+P338-P308+P313-P310-P501c

H318-H361f-H373

EINECSNumber : 208-591-9

RTECSNumber : FK9400000

TSCA : Yes

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures