Learn More
Cesium carbonate, 99.9% (metals basis)
Description
Adiponitrile is used as an intermediate for the synthesis of hexamethylenediamine (diamino-1,6-hexane). This chemical is an important intermediate for the manufacture of synthetic fiber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Melting Point | 610°C (decomposition) |
Physical Form | Powder/Granules |
Quantity | 25 g |
Assay Percent Range | 99.9% (metals basis) |
Beilstein | 4546405 |
Solubility Information | Freely soluble in water. Soluble in ethanol,dimethylformamide and dimethyl sulfoxide. |
IUPAC Name | dicaesium(1+) carbonate |
Formula Weight | 325.82 |
Sensitivity | Hygroscopic |
Density | 4.072 |
Show More |
Safety and Handling
Hazard Category | H318-H361f-H373 |
Precautionary Statement | P201-P202-P260-P281-P305+P351+P338-P308+P313-P310-P501c |
EINECSNumber | 208-591-9 |
RTECSNumber | FK9400000 |
TSCA | Yes |
storageNote1 | Hygroscopic |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.