Learn More
Cesium carbonate, Puratronic™, 99.994% (metals basis)
Description
Stainless Steel gauze is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Melting Point | 610°C (decomposition) |
Color | White |
Physical Form | Powder/Granules |
Quantity | 5 g |
Assay Percent Range | 99.994% (Metal basis) |
Beilstein | 4546405 |
Solubility Information | Highly soluble in water. Soluble in alcohol,dimethyl sulfoxide and N,N-dimethylformamide. Insoluble in toluene and p-xylene. |
IUPAC Name | dicesium;carbonate |
Formula Weight | 325.82 |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | H318-H361f-H373 |
Precautionary Statement | P201-P202-P260-P281-P305+P351+P338-P308+P313-P310-P501c |
EINECSNumber | 208-591-9 |
RTECSNumber | FK9400000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.