Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium chloride, 99.999% (metals basis)

Catalog No. aa1099314
Click to view available options
Quantity:
25 g
100 g

CAS: 7647-17-8 | ClCs | 168.36 g/mol

Cesium chloride is widely utilized as a precursor for the source of cesium ions. It finds an application in analytical chemistry and acts as a reagent to identify inorganic ions like aluminum, gallium and chromium by the color and morphology of the precipitate. Its radioisotopes are employed in nuclear medicine applications such as treatment of cancer and diagnosis of myocardial infarction. Occasionally, it can act as a phase transfer catalyst in selected organic reactions. It is applied in the manufacturing of electrical conducting glasses and screens of cathode ray tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1303°C
Melting Point 646°C
Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Soluble in water and concentrated hydrochloric acid.
IUPAC Name cesium;chloride
Formula Weight 168.36
Refractive Index 1.6418
Sensitivity Hygroscopic
Appearance White powder/crystalline/beads
Density 3.99
Chemical Name or Material Cesium chloride
Show More Show Less
Hazard Category H361-H373
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
EINECSNumber 231-600-2
RTECSNumber FK9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.