Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium fluoride, 99%, for analysis

Catalog No. p-4509657
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle

Catalog No. AC189510250

Supplier: Thermo Scientific Chemicals 189510250

Only null left
Add to Cart

CAS: 13400-13-0 | CsF | 151.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13400-13-0
Molecular Formula CsF
Molecular Weight (g/mol) 151.90
MDL Number MFCD00010960
InChI Key XJHCXCQVJFPJIK-UHFFFAOYSA-M
Synonym cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride
PubChem CID 25953
SMILES [F-].[Cs+]

Specifications

Boiling Point 1251.0°C
Melting Point 682.0°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 98.5% min
Linear Formula CsF
Fieser 01,121; 06,100; 07,57; 08,81; 09,100; 10,81; 11,115; 12,108; 13,68; 14,79; 15,75; 16,69; 17,68
Merck Index 15,2011
Solubility Information Solubility in water: 369g/100mL (18°C). Other solubilities: soluble in methanol, insoluble in dioxane and pyridine
IUPAC Name caesium(1+) fluoride
Formula Weight 151.91
Percent Purity 99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Cesium fluoride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
Toxic if inhaled.
Toxic if swallowed.
Suspected of damaging fertility or the unborn child if inhaled.
Contact with a
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISO
EINECSNumber 236-487-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.