Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium fluoride, Puratronic™, 99.99% (metals basis)

Catalog No. AA1212118
Click to view available options
Quantity:
1 g
10 g
50 g

Catalog No. AA1212118

Supplier: Thermo Scientific Chemicals 01212118

Only null left
Add to Cart

CAS: 13400-13-0 | CsF | 151.90 g/mol

Cesium fluoride is used for desilylation reactions in organic synthesis. Useful base in organic chemistry, due to its low nucleophilicity. Owing to its high ionic character, it is a very reactive source of fluoride ion in organic synthesis. Reagent of choice when reactive naked fluoride is required. Effective catalyst in promoting Knoevenegal condensation. Single crystals of CsF are useful as the windows of cells for infrared spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13400-13-0
Molecular Formula CsF
Molecular Weight (g/mol) 151.90
MDL Number MFCD00010960
InChI Key XJHCXCQVJFPJIK-UHFFFAOYSA-M
Synonym cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride
PubChem CID 25953
IUPAC Name cesium;fluoride
SMILES [F-].[Cs+]

Specifications

Boiling Point 1251°C
Melting Point 682°C
Color White
pH 6.5 to 7.5
Physical Form Crystalline
Quantity 50 g
UN Number UN2923
Solubility Information Soluble in water. Insoluble in dioxane and pyridine.
Formula Weight 151.9
Percent Purity 99.99%
Grade Puratronic
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cesium fluoride
Show More Show Less
Hazard Category H301+H311+H331-H315-H318-H361
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 236-487-3
RTECSNumber FK9650000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.