Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium formate hydrate, White chunks/powder, 99.9% (Metals basis), Thermo Scientific™

Catalog No. AA1292722
Click to view available options
Quantity:
100 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula CsOOCH·xH2O
MDL Number MFCD00039103

Specifications

Color White
Physical Form Chunks and/or Powder
Quantity 100 g
Assay Percent Range 99.9% (Metals basis)
Solubility Information Soluble in water.
Formula Weight 177.92 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Cesium formate hydrate
Hazard Category H315-H319
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 222-492-8
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.